missing translation for 'onlineSavingsMsg'
Learn More
alpha-Naphtholbenzein, Thermo Scientific Chemicals Product Code.: 11483157

alpha-Naphtholbenzein, Thermo Scientific Chemicals

Product Code. 11483157
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11483157

Brand: Thermo Scientific Alfa Aesar A16125.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).

Solubility
Soluble in ethanol, acetic acid and acetone. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, acids, water reactive substances, azo, diazo, hydrazines, metals, nitrides, peroxides, hydroperoxides, epoxides and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145-50-6
Molecular Formula C27H18O2
Molecular Weight (g/mol) 374.439
MDL Number MFCD00078492
InChI Key VDDWRTZCUJCDJM-PNHLSOANSA-N
PubChem CID 5941340
IUPAC Name (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one
SMILES C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O

Specifications

Melting Point ∼225°C (decomposition)
Quantity 5 g
Beilstein 3471575
Synonym p-naphtholbenzein, 1-naphtholbenzein, alpha-naphtholbenzein, .alpha.-naphtholbenzein, 4-4-hydroxynaphthyl phenylmethylene naphthalen-1-one, naphtholbenzein, 1 4h-naphthalenone, 4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone,4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone, 4-.alpha.-4-hydroxy-1-naphthyl benzylidene
Solubility Information Soluble in ethanol,acetic acid and acetone. Insoluble in water.
Formula Weight 374.44
Chemical Name or Material alpha-Naphtholbenzein

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249800 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberA16125.22,A16125.14,A16125.06
Lot Number10248281 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA16125.22,A16125.14,A16125.06
Lot Number10201226 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16125.22,A16125.14,A16125.06
Lot NumberF5244A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16125.22,A16125.14,A16125.06
Lot Number10100923 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16125.22,A16125.14,A16125.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
alpha-Naphtholbenzein, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.