missing translation for 'onlineSavingsMsg'
Learn More
Allyl alcohol, 99%, extra pure, Thermo Scientific Chemicals Product Code.: 11904621

Allyl alcohol, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 11904621
2.5 L, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11904621

Brand: Thermo Scientific Acros 102860025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-18-6
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00002920
InChI Key XXROGKLTLUQVRX-UHFFFAOYSA-N
Synonym allyl alcohol, 2-propen-1-ol, vinylcarbinol, 2-propenyl alcohol, 3-hydroxypropene, 2-propenol, allylic alcohol, 1-propen-3-ol, vinyl carbinol, weed drenchShow MoreShow Less
PubChem CID 7858
ChEBI CHEBI:16605
IUPAC Name prop-2-en-1-ol
SMILES OCC=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -129.0°C
Density 0.8500g/mL
Boiling Point 96.0°C to 98.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2OH
Refractive Index 1.4119 to 1.4139
Quantity 2.5 L
UN Number 1098
Beilstein 01,436
Merck Index 15,277
Specific Gravity 0.85
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, chloroform and ether
Formula Weight 58.08
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Allyl Alcohol, Extra Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0448600 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number102860010,102860025,102862500
Lot NumberA0448600 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number102860010,102860025,102862500
Lot NumberA0443061 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number102860010,102860025,102862500
Lot NumberA0398932 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number102860010,102860025,102862500
Lot NumberA0411898 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number102860010,102860025,102862500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl alcohol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Allyl alcohol, 99%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.