Learn More
Aalborg™ Air Mass Flowmeter
Brand:  Aalborg™ GFM17A-VAL6-A0/3AB-10-SS
 View more versions of this product
Product Code. 11892170
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Specifications
Mass Flowmeter
0 to 10 LPM
12 V to 200 mA max
Full scale
±5%
NIST calibration certificate
±1.5 %
5°C to 50°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products