missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aesculin Blood Agar (Modified)

Isolate and differentiate bacteria involved in bovine mastitis with Thermo Scientific™ Aesculin Blood Agar prepared media plates.

Brand:  Thermo Scientific™ PB5023A

12.55 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10669244

  • 18.20€ / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Isolate and differentiate bacteria involved in bovine mastitis with Thermo Scientific™ Aesculin Blood Agar (Modified). Due to the relatively high agar content, the medium exhibits a high gel strength so that all samples of a quarter milk can be spread onto a single plate for greater efficiency. The inclusion of blood in the medium ensures the growth of staphylococci and streptococci and permits direct detection of enterococci and S.uberis by means of aesculin cleaving.

Mastitis in cattle causes large losses in dairy farming. After fertility disorders, udder disorders are the second most important cause of death among dairy cattle. The bacteriological investigation of milk samples is the most important diagnostic measure in the detection of udder disorders. Aesculin Blood Agar supports the isolation of most the frequent bacterial pathogens associated with mastitis.
  • Supports the growth of staphylococci and streptococci
  • Permits direct detection of enterococci and S.uberis
  • Has the ready-to-use convenience of prepared media
  • Allows all samples of a quarter milk to be spread on a single plate
Over 130 different microorganisms have been isolated from milk samples in connection with mastitis among cattle; however, a relatively narrow spectrum of bacteria number among the most frequent bacterial pathogens: Staphylococcus aureus, Streptococcus agalactiae, S. dysgalactiae and S. uberi, as well as other streptococci and coliform bacteria.

Due to the inclusion of blood, Aesculin Blood Agar ensures the growth of staphylococci and streptococci, and permits direct detection of enterococci and S.uberis by means of aesculin cleaving. The growth of coliform bacteria, pseudomonads and yeasts is also possible on the non-selective agar. In comparison with coliform bacteria, streptococci as well as yeasts grow in smaller colonies than, for example, on Columbia agar with sheep blood.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

6°C to 12°C
Agar
Wrapped in single cellulose-based film wrap
90mm plate
17.5g ± 5%
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6260887 Certificate TypeCertificate of Analysis Date23/04/2025 Catalog NumberPB5023A
Lot Number6254986 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberPB5023A
Lot Number6249104 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog NumberPB5023A
Lot Number6242416 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog NumberPB5023A
Lot Number6239735 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberPB5023A
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Aesculin Blood Agar (Modified) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.