missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Esculape Laboratoire™ ADR First Aid Kit
Brand: Esculape Laboratoire™ 7220305
This item is not returnable.
View return policy
Specifications
Transporting hazardous materials with eye washing 200mL | |
ADR First Aid Kit | |
1 200mL eyewash with eye solution, 1 survival blanket, 1 hemostatic pad, 1 Cotton bag 25 g, 2 bags of 5 sterile compresses 20 x 20cm, 2 gauze bandages 3mx 7cm, 1 bag of 10 dressings, 2 Plastic bottles 125mL, 1 pair of scissors, 1 splinter tweezers, 1 fing | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Esculape Laboratoire™ ADR First Aid Kit > For Use With Transporting hazardous materials with eye washing 200mL
Spot an opportunity for improvement?Share a Content Correction