Learn More
Shamrock™ Adhesive Autoclave Indicator Tape
Indicates completion of normal steam sterilization processing. Shamrock™ Adhesive Autoclave Indicator Tape is versatile and accepts handwritten notations - may be used to seal and identify laboratory materials before and after processing.
Brand: Shamrock™ SIL-10-1
Description
- Resistant to water, oil and acid
- “Sterilized” appears after reaching 121°C
- For use in autoclaves
Specifications
For the sterilization in Poupinel or in an autoclave | |
12 m | |
121°C | |
1 Ea. | |
15 min. |
Poupinel or in an autoclave | |
Adhesive Autoclave Indicator Tape | |
With indicator, Resistant to water, oil and acid | |
Yes | |
Poupinel or in an autoclave |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.