Learn More
Memmert™ Additional Reinforced Stainless Steel Shelf (For Reinforced Chamber Option), For Ich750
Brand: Memmert™ ICH750B32191
Includes: Guide Bars And Fixing Screws
Specifications
Reinforced Shelf | |
Only in connection with option K1 | |
60 kg |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Memmert™ Additional Reinforced Stainless Steel Shelf (For Reinforced Chamber Option), For Ich750 >