Learn More
Memmert™ Additional Reinforced Stainless Steel Grid (For Reinforced Chamber Option), For Ich750
Use these grids with Memmert ovens. Memmert™ Reinforced Grids are designed to hold up to 60 kg.
Brand:  Memmert™ ICH750B32190
Product Code. 16336687
  • 635.00€ / Each
Estimated Shipment: 11-07-2025
to see stock.
Explore more special offers
Add to basket
Includes: Guide Bars And Fixing Screws
Description
Description
  • Reinforced stainless steel grid
  • Max. loading 60 kg
  • From size 450 with guide bars and fixing screws and only in connection with option K1
Specifications
Specifications
Reinforced Grid
Only in connection with option K1
60 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Additional Reinforced Stainless Steel Grid (For Reinforced Chamber Option), For Ich750 >