missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetoacetyl CoA synthetase Antibody (011), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89801PE
This item is not returnable.
View return policy
Description
Acetoacetyl CoA synthetase Monoclonal antibody specifically detects Acetoacetyl CoA synthetase in Human samples. It is validated for ELISA
Specifications
Acetoacetyl CoA synthetase | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
65985 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
011 | |
ELISA | |
acetoacetate-CoA ligase, acetoacetyl-CoA synthetase, ACSF1FLJ41251, Acyl-CoA synthetase family member 1, EC 6.2.1, EC 6.2.1.16, FLJ12389, homolog of C. elegans supressor of ras 5 (sur-5), Protein sur-5 homolog, SUR-5 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Acetoacetyl CoA synthetase (Accession#: NP_076417.2; Met1-Phe672). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetoacetyl CoA synthetase Antibody (011), PE, Novus Biologicals™ > 0.1 mL; PE
Spot an opportunity for improvement?Share a Content Correction