missing translation for 'onlineSavingsMsg'
Learn More
Acetic Acid Solution, 0.1M, Chem-Lab Product Code.: 11903103

Acetic Acid Solution, 0.1M, Chem-Lab

Product Code. 11903103
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 11903103

Brand: Chem Lab CL05.0107.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for laboratory use in titrations, as a chemical reagent and as a buffer. Acetic Acid Solution, 0.1M, Chem-Lab is a clear, colorless solution with a vinegar odor. It contains 6.005g CH3COOH per 1L H2O.

Acetic acid is a clear, colorless solution with a strong sour odor. It is the second simplest carboxylic acid (after formic acid) and consists of two small functional groups, an acetyl group and a hydroxyl group. Undiluted, it is referred to as “glacial” acetic acid.

Acetic acid is an important chemical reagent and industrial chemical for the production of chemical compounds. It is used in the production of photographic film, adhesives, paints, and synthetic fibers and fabrics. It is a primary component in the production of vinyl acetate monomer, as well as acetic anhydride and ester production. The volume of acetic acid used in vinegar is comparatively small.

Specifications

pH <4
CAS 64-19-7, 7732-18-5
Color Colorless
Molecular Formula C2H4O2
MDL Number MFCD00036152
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol
Solubility Information Soluble in water
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
Concentration 0.1M
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
PubChem CID 176
ChEBI CHEBI:15366
Formula Weight 60.05g/mol
Density 1.00g/mL
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetic Acid Solution, 0.1M, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.