Learn More
Thermo Scientific™ Accessory Base for Programmable Electronic High-Power Crimp Stations
Free hands for faster crimping operations with an adjustable base.
Brand: Thermo Scientific™ ECRH-STAND
223.53 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Free hands for faster crimping operations with the adjustable Thermo Scientific™ Accessory Base for Programmable Electronic High-Power Crimp Stations.
- Adjustable height with solvent-resistant base plate
- Dimensions: 14.8 x 10 x 7″ (37.6 x 25.4 x 17.7cm)
Specifications
Accessory Base for Electronic Crimps (base plate and bar) | |
1 Ea. | |
Electronic Crimp Station |
14.8 x 10 x 7 in. | |
Stand | |
Industrial Chromatography |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.