Learn More
Fisherbrand™ Accessories for Incubating Mini Shaker
Accessories required for the use of Fisher Scientific™ Incubating Mini Shaker
Brand: Fisherbrand™ 88861144
34.25 EUR valid until 2025-03-31
Use promo code "24162" to get your promotional price.
Description
Accessories for incubating mini shaker
- Increase performance and flexibility
Specifications
140 x 140mm Sticky Mat |
Promotions
-
Multiple Manufacturers
Consumables Monthly Promotion
Explore exclusive monthly deals on essential consumables for unbeatable savings.Promo Code: 24222Expires: 2025-03-31
New Offer Save Up to 92% -
Fisherbrand
Fisherbrand Sweet Deals on Top Products
Get up to 60% off select Fisherbrand productsPromo Code: 24162Expires: 2025-03-31