Learn More
Fisherbrand™ Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers
Optimize performance and flexibility
Brand:  Fisherbrand™ 88860143
30.06 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 15386607
  • 35.60€ / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers. Optimize performance and flexibility
  • Accessory for use with RT Hotplate, RT Basic and Advanced Hotplate Stirrers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For use with RT Hotplate, RT Basic and Advanced Hotplate Stirrers
Support Rod (12mm dia., 400mm, M10)
Promotions
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24163
    Expires2025-04-30
    New Offer
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/04/2025 and 30/04/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
Product Content Correction
Product Title
Fisherbrand™ Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers > Support Rod (12mm dia., 400mm, M10)