missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AC 5216
Selective TSPO ligand
243.00€ - 956.00€
Specifications
Purity | >98% |
---|---|
Product Type | AC 5216 |
Molecular Weight (g/mol) | 401.46 |
Inhibitors | AC 5216 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18756218
|
Tocris Bioscience™
5281/10 |
10 mg |
243.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18789806
|
Tocris Bioscience™
5281/50 |
50 mg |
956.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ligand of translocator protein (TSPO, peripheral benzodiazepine receptor). Exhibits high affinity for TSPO in rat whole brain (Ki = 0.297 nM), and high potency against human and rat glial TSPO (IC50 values are 2.73 and 3.04 nM respectively). Displays no noticeable binding to a number of other receptors, transporters or ion channels. Also displays anxiolytic and antidepressant effects in rodent models, without inducing benzodiazepine-like adverse effects.Specifications
>98% | |
AC 5216 | |
401.46 | |
AC 5216 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title