missing translation for 'onlineSavingsMsg'
Learn More
A 784168, Tocris Bioscience™ Product Code.: 18085072

A 784168, Tocris Bioscience™

Product Code. 18085072
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18085072

Brand: Tocris Bioscience 4319/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective TRPV1 antagonist

Potent TRPV1 antagonist (IC50 = 25 nM for inhibition of TRPV1 activation by 50 nM capsaicin). Displays no activity against a range of receptors, including TRPA1, GABA, opioid, and purinergic receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824982-41-4
Molecular Formula C19H15F6N3O3S
Molecular Weight (g/mol) 479.397
InChI Key SDUAWRFBHRAFBM-UHFFFAOYSA-N
Synonym unii-lb160j37hd, 3'-trifluoromethyl-n-4-trifluoromethyl sulfonyl phenyl-3,6-dihydro-2h-1,2'-bipyridine-4-carboxamide, n-4-trifluoromethanesulfonylphenyl-3'-trifluoromethyl-3,6-dihydro-2h-1,2'-bipyridine-4-carboxamide, 1 2h ,2'-bipyridine-4-carboxamide, 3,6-dihydro-3'-trifluoromethyl-n-4-trifluoromethyl sulfonyl phenyl, 1-3-trifluoromethyl pyridin-2-yl-n-4-trifluoromethylsulfonyl phenyl-1,2,3,6-tetrahydropyridine-4-carboxamide, 3,6-dihydro-3'-trifluoromethyl-n-4-trifluoromethyl sulfonyl phenyl-1 2h ,2'-bipyridine-4-carboxamide, n-4-trifluoromethane sulfonylphenyl-1-3-trifluoromethyl pyridin-2-yl-1,2,3,6-tetrahydropyridine-4-carboxamide
PubChem CID 11420211
IUPAC Name 1-[3-(trifluoromethyl)pyridin-2-yl]-N-[4-(trifluoromethylsulfonyl)phenyl]-3,6-dihydro-2H-pyridine-4-carboxamide
SMILES C1CN(CC=C1C(=O)NC2=CC=C(C=C2)S(=O)(=O)C(F)(F)F)C3=C(C=CC=N3)C(F)(F)F

Specifications

Quantity 50 mg
Formula Weight 479.4
Percent Purity >99%
Chemical Name or Material A 784168
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
A 784168, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.