missing translation for 'onlineSavingsMsg'
Learn More
8-Hydroxyquinoline, 99%, Thermo Scientific Chemicals Product Code.: 11457746

8-Hydroxyquinoline, 99%, Thermo Scientific Chemicals

Product Code. 11457746
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11457746

Brand: Thermo Scientific Alfa Aesar A14720.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

8-Hydroxyquinoline is used as an antiseptic and disinfectant. Its alcoholic solution is used in liquid bandages. It is used to prepare tris(8-hydroxyquinolinato)aluminum, which finds application as a component of organic light-emitting diodes (OLED's). It is also used as a reagent and as a stabilizer of hydrogen peroxide in a rocket fuel oxidizer. It is used as a chelating agent and used as a carrier for radio-indium for diagnostic purposes. Its halogenated derivatives are used as anti-infective agents and oral antiamebics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Hydroxyquinoline is used as an antiseptic and disinfectant. Its alcoholic solution is used in liquid bandages. It is used to prepare tris(8-hydroxyquinolinato)aluminum, which finds application as a component of organic light-emitting diodes (OLED′s). It is also used as a reagent and as a stabilizer of hydrogen peroxide in a rocket fuel oxidizer. It is used as a chelating agent and used as a carrier for radio-indium for diagnostic purposes. Its halogenated derivatives are used as anti-infective agents and oral antiamebics.

Solubility
Almost insoluble in water and diethyl ether. Highly soluble in ethanol, acetone, chloroform, and benzene.Soluble in acetic acid, ethanol, acetone, chloroform, benzene and aqueous mineral acids. Insoluble in water and diethylether.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148-24-3
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006807
InChI Key MCJGNVYPOGVAJF-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
PubChem CID 1923
ChEBI CHEBI:48981
IUPAC Name quinolin-8-ol
SMILES OC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point 72°C to 76°C
Boiling Point 267°C
Quantity 500 g
Beilstein 114512
Sensitivity Light sensitive
Merck Index 14,4843
Solubility Information Almost insoluble in water and diethyl ether; Highly soluble in ethanol,acetone,chloroform,and benzene;Soluble in acetic acid,ethanol,acetone,chloroform,benzene and aqueous mineral acids; Insoluble in water and diethylether.
Formula Weight 145.16
Percent Purity 99%
Chemical Name or Material 8-Hydroxyquinoline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252579 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog NumberA14720.22,A14720.0E,A14720.36
Lot Number10245296 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog NumberA14720.22,A14720.0E,A14720.36
Lot Number10242650 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog NumberA14720.22,A14720.0E,A14720.36
Lot Number10238376 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14720.22,A14720.0E,A14720.36
Lot Number10060763 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14720.22,A14720.0E,A14720.36
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 8-Hydroxyquinoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H360-May damage fertility or the unborn child.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 8-Hydroxyquinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
8-Hydroxyquinoline, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.