Learn More
5-Nitrothiophene-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
78.79€ - 386.93€
Chemical Identifiers
CAS | 4521-33-9 |
---|---|
Molecular Formula | C5H3NO3S |
Molecular Weight (g/mol) | 157.143 |
MDL Number | MFCD00005433 |
InChI Key | CHTSWZNXEKOLPM-UHFFFAOYSA-N |
Synonym | 5-nitrothiophene-2-carboxaldehyde, 5-nitro-2-thiophenecarboxaldehyde, 5-nitro-2-thiophenecarbaldehyde, 5-nitro-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-nitro, 5-nitrothiophene-2-aldehyde, thiophenecarboxaldehyde, 5-nitro, pubchem15668, 5-nitrothiophen aldehyde, 5-nitro-2-formyl-thiopheneShow More |
PubChem CID | 78281 |
IUPAC Name | 5-nitrothiophene-2-carbaldehyde |
SMILES | C1=C(SC(=C1)[N+](=O)[O-])C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371297
|
Thermo Scientific Alfa Aesar
L01493.06 |
5 g |
78.79€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381297
|
Thermo Scientific Alfa Aesar
L01493.14 |
25 g |
386.93€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.
Solubility
Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
4521-33-9 | |
157.143 | |
CHTSWZNXEKOLPM-UHFFFAOYSA-N | |
78281 | |
C1=C(SC(=C1)[N+](=O)[O-])C=O |
C5H3NO3S | |
MFCD00005433 | |
5-nitrothiophene-2-carboxaldehyde, 5-nitro-2-thiophenecarboxaldehyde, 5-nitro-2-thiophenecarbaldehyde, 5-nitro-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-nitro, 5-nitrothiophene-2-aldehyde, thiophenecarboxaldehyde, 5-nitro, pubchem15668, 5-nitrothiophen aldehyde, 5-nitro-2-formyl-thiopheneShow More | |
5-nitrothiophene-2-carbaldehyde |
Specifications
4521-33-9 | |
C5H3NO3S | |
120540 | |
5-nitrothiophene-2-carboxaldehyde, 5-nitro-2-thiophenecarboxaldehyde, 5-nitro-2-thiophenecarbaldehyde, 5-nitro-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-nitro, 5-nitrothiophene-2-aldehyde, thiophenecarboxaldehyde, 5-nitro, pubchem15668, 5-nitrothiophen aldehyde, 5-nitro-2-formyl-thiophene | |
CHTSWZNXEKOLPM-UHFFFAOYSA-N | |
5-nitrothiophene-2-carbaldehyde | |
78281 | |
98% |
72°C to 76°C | |
MFCD00005433 | |
Air sensitive | |
Insoluble in water. | |
C1=C(SC(=C1)[N+](=O)[O-])C=O | |
157.143 | |
157.15 | |
5-Nitrothiophene-2-carboxaldehyde |
Safety and Handling
missing translation for 'einecsNumber' : 224-850-9
missing translation for 'tsca' : No
Recommended Storage : Keep cold