missing translation for 'onlineSavingsMsg'
Learn More
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals Product Code.: 11430458

5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals

Product Code. 11430458
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11430458

Brand: Thermo Scientific Alfa Aesar B24601.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1046-56-6
Molecular Formula C20H14N4
Molecular Weight (g/mol) 310.36
MDL Number MFCD00006462
InChI Key OTMYLOBWDNFTLO-UHFFFAOYSA-N
Synonym 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine, 5,6-diphenyl-3-2-pyridyl-1,2,4-triazine, 1,2,4-triazine, 5,6-diphenyl-3-2-pyridinyl, 5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine, as-triazine, 5,6-diphenyl-3-2-pyridyl, 5,6-diphenyl-3-2-pyridinyl-1,2,4-triazine, maybridge1_007490, bio7c7, acmc-1boy4, cbmicro_025706
PubChem CID 70588
SMILES C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Melting Point 189°C to 192°C
Quantity 5 g
Beilstein 542151
Solubility Information Soluble in water.
IUPAC Name 5,6-diphenyl-3-(pyridin-2-yl)-1,2,4-triazine
Formula Weight 310.36
Percent Purity 99%
Chemical Name or Material 5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine
compliance-icons
Product Identifier
  • 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.