Learn More
4-Nitrobenzoic acid, 99%, Thermo Scientific Chemicals
Description
4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.
Solubility
Very slightly soluble in cold water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Specifications
Specifications
Melting Point | 239°C to 241°C |
Density | 1.61 |
Quantity | 1000 g |
Beilstein | 973593 |
Merck Index | 14,6589 |
Solubility Information | Very slightly soluble in cold water. |
Formula Weight | 167.12 |
Percent Purity | 99% |
Chemical Name or Material | 4-Nitrobenzoic acid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10243554 | Certificate of Analysis | 13/10/2022 | A14738.30,A14738.0B,A14738.0I |
J4057B | Certificate of Analysis | 22/11/2021 | A14738.30,A14738.0B,A14738.0I |
10141876 | Certificate of Analysis | 22/11/2021 | A14738.30,A14738.0B,A14738.0I |
J19Y057 | Certificate of Analysis | 22/11/2021 | A14738.30,A14738.0B,A14738.0I |
10222151 | Certificate of Analysis | 22/11/2021 | A14738.30,A14738.0B,A14738.0I |
Safety and Handling


- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.