missing translation for 'onlineSavingsMsg'
Learn More
4-Methylbenzenesulphonylhydrazide, 97%, Thermo Scientific Chemicals Product Code.: 11415643

4-Methylbenzenesulphonylhydrazide, 97%, Thermo Scientific Chemicals

Product Code. 11415643
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11415643

Brand: Thermo Scientific Alfa Aesar A13529.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

p-Toluenesulfonyl hydrazide is used as a reagent for the preparation of tosylhydrazones. It was used to prepare dipyrazolo[1,5-a:4,3-c]pyridines and 1,2,3-selenadiazole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonyl hydrazide is used as a reagent for the preparation of tosylhydrazones. It was used to prepare dipyrazolo[1,5-a:4′,3′-c]pyridines and 1,2,3-selenadiazole derivatives.

Solubility
Soluble in water, alcohol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1576-35-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00007588
InChI Key ICGLPKIVTVWCFT-UHFFFAOYSA-N
Synonym 4-methylbenzenesulfonhydrazide, p-toluenesulfonhydrazide, tosylhydrazine, p-toluenesulfonyl hydrazide, tosylhydrazide, p-tosylhydrazine, p-toluenesulfonylhydrazine, celogen tsh, ptsh, 4-toluenesulfonyl hydrazideShow MoreShow Less
PubChem CID 15303
IUPAC Name 4-methylbenzenesulfonohydrazide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN

Specifications

Melting Point 108°C to 111°C
Density 1.4
Quantity 500 g
UN Number UN3226
Beilstein 610130
Solubility Information Soluble in water,alcohol.
Formula Weight 186.23
Percent Purity 98%
Chemical Name or Material p-Toluenesulfonyl hydrazide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252669 Certificate TypeCertificate of Analysis Date27/11/2024 Catalog NumberA13529.22,A13529.14,A13529.36
Lot Number10251846 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog NumberA13529.22,A13529.14,A13529.36
Lot Number10240427 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberA13529.22,A13529.14,A13529.36
Lot Number10240428 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberA13529.22,A13529.14,A13529.36
Lot Number10241075 Certificate TypeCertificate of Analysis Date08/06/2022 Catalog NumberA13529.22,A13529.14,A13529.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methylbenzenesulphonylhydrazide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.