missing translation for 'onlineSavingsMsg'
Learn More
4-Formylphenoxyacetic acid, 98%, Thermo Scientific Chemicals Product Code.: 11451708

4-Formylphenoxyacetic acid, 98%, Thermo Scientific Chemicals

Product Code. 11451708
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11451708

Brand: Thermo Scientific Alfa Aesar A13960.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Formylphenoxyacetic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Formylphenoxyacetic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in hot methanol within almost transparency.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22042-71-3
Molecular Formula C9H7O4
Molecular Weight (g/mol) 179.15
MDL Number MFCD00016613
InChI Key OYNIIKHNXNPSAG-UHFFFAOYSA-M
Synonym 4-formylphenoxyacetic acid, 4-formylphenoxy acetic acid, 2-4-formylphenoxy acetic acid, p-formylphenoxyacetic acid, acetic acid, 4-formylphenoxy, acetic acid, 2-4-formylphenoxy, p-formylphenoxy acetic acid, 4-carboxymethyloxy benzaldehyde, 4-formyl phenoxy acetic acid, 4-formylphenoxyactic acidShow MoreShow Less
PubChem CID 89177
IUPAC Name 2-(4-formylphenoxy)acetic acid
SMILES [O-]C(=O)COC1=CC=C(C=O)C=C1

Specifications

Melting Point 193°C to 198°C
Quantity 50 g
Beilstein 779885
Sensitivity Air Sensitive
Solubility Information Soluble in hot methanol within almost transparency.
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 4-Formylphenoxyacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10190597 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog NumberA13960.18
Lot NumberGBFA003216 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13960.30,A13960.09,A13960.18
Lot NumberH6993A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13960.30,A13960.09,A13960.18
Lot Number10190597 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13960.30,A13960.09,A13960.18
Lot NumberF2634C Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13960.30,A13960.09,A13960.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Formylphenoxyacetic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.