missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-N-Boc-L-phenylalanine, 95%, Thermo Scientific Chemicals
46.75€ - 439.00€
Chemical Identifiers
CAS | 55533-24-9 |
---|---|
Molecular Formula | C14H20N2O4 |
Molecular Weight (g/mol) | 280.32 |
MDL Number | MFCD00038139 |
InChI Key | NDMVQEZKACRLDP-NSHDSACASA-N |
Synonym | boc-phe 4-nh2-oh, boc-4-amino-l-phenylalanine, boc-l-4-aminophenylalanine, 4-amino-n-boc-l-phenylalanine, s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 4-amino-n-tert-butoxycarbonyl-l-phenylalanine, 2s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-4-aminophenyl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-4-aminophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid |
PubChem CID | 2761482 |
IUPAC Name | (2S)-3-(4-aminophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(N)C=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15498457
|
Thermo Scientific Alfa Aesar
H51980.MD |
250 mg |
46.75€
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15478457
|
Thermo Scientific Alfa Aesar
H51980.03 |
1 g |
125.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15488457
|
Thermo Scientific Alfa Aesar
H51980.06 |
5 g |
439.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
55533-24-9 | |
280.32 | |
NDMVQEZKACRLDP-NSHDSACASA-N | |
2761482 | |
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(N)C=C1)C(O)=O |
C14H20N2O4 | |
MFCD00038139 | |
boc-phe 4-nh2-oh, boc-4-amino-l-phenylalanine, boc-l-4-aminophenylalanine, 4-amino-n-boc-l-phenylalanine, s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 4-amino-n-tert-butoxycarbonyl-l-phenylalanine, 2s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-4-aminophenyl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-4-aminophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid | |
(2S)-3-(4-aminophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
55533-24-9 | |
C14H20N2O4 | |
boc-phe 4-nh2-oh, boc-4-amino-l-phenylalanine, boc-l-4-aminophenylalanine, 4-amino-n-boc-l-phenylalanine, s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 4-amino-n-tert-butoxycarbonyl-l-phenylalanine, 2s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-4-aminophenyl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-4-aminophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid | |
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(N)C=C1)C(O)=O | |
280.32 | |
280.32 | |
4-Amino-N-Boc-L-phenylalanine |
124°C to 126°C | |
MFCD00038139 | |
NDMVQEZKACRLDP-NSHDSACASA-N | |
(2S)-3-(4-aminophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2761482 | |
95% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only