missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-N-Boc-L-phenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15488457
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55533-24-9
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00038139
InChI Key NDMVQEZKACRLDP-NSHDSACASA-N
Synonym boc-phe 4-nh2-oh, boc-4-amino-l-phenylalanine, boc-l-4-aminophenylalanine, 4-amino-n-boc-l-phenylalanine, s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 4-amino-n-tert-butoxycarbonyl-l-phenylalanine, 2s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-4-aminophenyl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-4-aminophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid
PubChem CID 2761482
IUPAC Name (2S)-3-(4-aminophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(N)C=C1)C(O)=O

Specifications

Melting Point 124°C to 126°C
Quantity 5 g
Formula Weight 280.32
Percent Purity 95%
Chemical Name or Material 4-Amino-N-Boc-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'