missing translation for 'onlineSavingsMsg'
Learn More
4,6-Dihydroxy-2-mercaptopyrimidine, 98%, Thermo Scientific Chemicals Product Code.: 11410573

4,6-Dihydroxy-2-mercaptopyrimidine, 98%, Thermo Scientific Chemicals

Product Code. 11410573
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11410573

Brand: Thermo Scientific Alfa Aesar A12681.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4,6-Dihydroxy-2-mercaptopyrimidine is used as a reagent in assaying malondialdehyde (theTBARS assay of lipid peroxidation). It is used to detect lipid oxidation, quantitating sialic acids and lipopolysaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-17-6
Molecular Formula C4H4N2O2S
Molecular Weight (g/mol) 144.15
MDL Number MFCD00006674
InChI Key RVBUGGBMJDPOST-UHFFFAOYSA-N
Synonym 2-thiobarbituric acid, thiobarbituric acid, bathyran, austranal, 4,6-dihydroxy-2-thiopyrimidine, 2-thioxodihydropyrimidine-4,6 1h,5h-dione, 2-thio-4,6-dioxypyrimidine, usaf ek-660, 4,6 1h,5h-pyrimidinedione, dihydro-2-thioxo, 2-mercaptobarbituric acidShow MoreShow Less
PubChem CID 2723628
ChEBI CHEBI:33202
IUPAC Name 2-sulfanylidene-1,3-diazinane-4,6-dione
SMILES O=C1CC(=O)NC(=S)N1

Specifications

Melting Point >300°C
pH 1.8
Quantity 100 g
Beilstein 120663
Formula Weight 144.15
Percent Purity 98%
Chemical Name or Material 4,6-Dihydroxy-2-mercaptopyrimidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250376 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberA12681.22,A12681.14,A12681.36
Lot Number10247925 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog NumberA12681.22,A12681.14,A12681.36
Lot NumberK17X034 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12681.22,A12681.14,A12681.36
Lot NumberUSLF000585 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12681.22,A12681.14,A12681.36
Lot NumberG25S001 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12681.22,A12681.14,A12681.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,6-Dihydroxy-2-mercaptopyrimidine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.