Learn More
Thermo Scientific™ 3K/4K Series Particle Counter Standards Product Code.: 11894992
  • ONLINE EXCLUSIVE PRICE
    357.00€
    201.85€ / 15mL
    Save 155.15€
    43% Off
Thermo Scientific™ 3K/4K Series Particle Counter Standards
Product Code. 11894992 Shop All Thermo Scientific Products
0.15 μm, 15mL
Click to view available options
Diameter (Metric):
0.1 μm
0.15 μm
0.2 μm
0.22 μm
0.27 μm
0.3 μm
0.35 μm
0.4 μm
0.5 μm
0.6 μm
0.7 μm
0.8 μm
0.9 μm
0.99 μm
1.0 μm
1.6 μm
2.0 μm
3 μm
4 μm
5.0 μm
6 μm
7 μm
10 μm
15 μm
20 μm
25 μm
30 μm
40 μm
50 μm
60 μm
70 μm
80 μm
100 μm
Unit Size:
15mL
Pack of 30
Pack of 50
Pack of 80
Product Code. 11894992
Brand: Thermo Scientific™ 3K150
  • ONLINE EXCLUSIVE PRICE
    357.00€
    201.85€ / 15mL
    Save 155.15€
    43% Off
Ensure accurate and convenient calibration of laser particle counters with Thermo Scientific™ 3K/4K Series Particle Counter Standards.
Ensure accurate and convenient calibration of laser particle counters with Thermo Scientific™ 3K/4K Series Particle Counter Standards. Compared with our 3000 and 4000 Series products, this product line is precisely diluted for immediate use in laser particle counters with minimal time-consuming concentration adjustments.
Features:
  • Suspensions of monodisperse polystyrene spheres for use in airborne or liquid particle dispersion systems
  • Particle diameters traceable to the Standard Meter through the National Institute of Standards and Technology (NIST)
  • Accurately calibrates or checks performance of laser particle counters used in cleanrooms and other contamination monitoring applications
  • Particles prepared as low residue aqueous suspensions for minimal background interference
TRUSTED_SUSTAINABILITY
Specifications
Additive Contains trace amount of surfactant to inhibit agglomeration and promote stability
Concentration 1 x 109/mL
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Contents Polymer particles in water
Description NIST traceable particles that provide calibration of laser particle counters
Color White or Colorless
For Use With (Application) Laser Particle Counter Calibration
Material Polystyrene
Product Type Count Control Particles
Includes Certificate of Calibration and Traceability to NIST
Particle Density 1.05g/cm3
Quantity 15 mL
Refractive Index 1.59 at 589nm (25°C)
Size Distribution ≤ 2%
Stability Product stable for a minimum of 12 months
Diameter (Metric) 0.15 μm
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number255495 Certificate TypeCertificate of Analysis Date21/02/2025 Product Code3K-150
Lot Number269438 Certificate TypeCertificate of Analysis Date21/02/2025 Product Code3K-150
Lot Number251077 Certificate TypeCertificate of Analysis Date21/02/2025 Product Code3K-150
Lot Number258899 Certificate TypeCertificate of Analysis Date21/02/2025 Product Code3K-150
Lot Number281314_0526 Certificate TypeCertificate of Analysis Date21/02/2025 Product Code3K-150
Product Content Correction
Product Title
Thermo Scientific™ 3K/4K Series Particle Counter Standards >