Learn More
Thermo Scientific™ 3K/4K Series Particle Counter Standards
Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
0.1 μm
0.15 μm
0.2 μm
0.22 μm
0.27 μm
0.3 μm
0.35 μm
0.4 μm
0.5 μm
0.6 μm
0.7 μm
0.8 μm
0.9 μm
0.99 μm
1.0 μm
1.6 μm
2.0 μm
3 μm
4 μm
5.0 μm
6 μm
7 μm
10 μm
15 μm
20 μm
25 μm
30 μm
40 μm
50 μm
60 μm
70 μm
80 μm
100 μm
Unit Size:
15mL
Pack of 30
Pack of 50
Pack of 80
Description
Ensure accurate and convenient calibration of laser particle counters with Thermo Scientific™ 3K/4K Series Particle Counter Standards. Compared with our 3000 and 4000 Series products, this product line is precisely diluted for immediate use in laser particle counters with minimal time-consuming concentration adjustments.
Features:
- Suspensions of monodisperse polystyrene spheres for use in airborne or liquid particle dispersion systems
- Particle diameters traceable to the Standard Meter through the National Institute of Standards and Technology (NIST)
- Accurately calibrates or checks performance of laser particle counters used in cleanrooms and other contamination monitoring applications
- Particles prepared as low residue aqueous suspensions for minimal background interference
Specifications
Specifications
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 3 x 104/mL |
Content And Storage | Unless otherwise stated, refrigerate 2° to 8deg;C product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. |
Contents | Polymer particles in water |
Description | NIST traceable particles that provide calibration of laser particle counters |
Color | White or Colorless |
For Use With (Application) | Laser Particle Counter Calibration |
Material | Polystyrene |
Product Type | Count Control Particles |
Includes | Certificate of Calibration and Traceability to NIST |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
287871_0626 | Certificate of Analysis | 21/02/2025 | 4K-80 |
270368 | Certificate of Analysis | 21/02/2025 | 4K-80 |
256299 | Certificate of Analysis | 21/02/2025 | 4K-80 |
260831 | Certificate of Analysis | 21/02/2025 | 4K-80 |
278525_0925 | Certificate of Analysis | 21/02/2025 | 4K-80 |
Product Content Correction
Product Title
Thermo Scientific™ 3K/4K Series Particle Counter Standards >