Learn More
Thermo Scientific™ 3K/4K Series Particle Counter Standards
Shop All Thermo Scientific ProductsDescription
Ensure accurate and convenient calibration of laser particle counters with Thermo Scientific™ 3K/4K Series Particle Counter Standards. Compared with our 3000 and 4000 Series products, this product line is precisely diluted for immediate use in laser particle counters with minimal time-consuming concentration adjustments.
- Suspensions of monodisperse polystyrene spheres for use in airborne or liquid particle dispersion systems
- Particle diameters traceable to the Standard Meter through the National Institute of Standards and Technology (NIST)
- Accurately calibrates or checks performance of laser particle counters used in cleanrooms and other contamination monitoring applications
- Particles prepared as low residue aqueous suspensions for minimal background interference
Specifications
Specifications
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 1 x 107/mL |
Content And Storage | Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. |
Contents | Polymer particles in water |
Description | NIST traceable particles that provide calibration of laser particle counters |
Color | White or Colorless |
For Use With (Application) | Laser Particle Counter Calibration |
Material | Polystyrene |
Product Type | Count Control Particles |
Includes | Certificate of Calibration and Traceability to NIST |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
272503 | Certificate of Analysis | 21/02/2025 | 4K-06 |
278516_0925 | Certificate of Analysis | 21/02/2025 | 4K-06 |
259784 | Certificate of Analysis | 21/02/2025 | 4K-06 |
252362 | Certificate of Analysis | 05/04/2023 | 4K-06 |
221781 | Certificate of Analysis | 28/01/2022 | 4K-06 |