missing translation for 'onlineSavingsMsg'
Learn More
3-Phenoxyaniline, 98%, Thermo Scientific Chemicals Product Code.: 11428107

3-Phenoxyaniline, 98%, Thermo Scientific Chemicals

Product Code. 11428107
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11428107

Brand: Thermo Scientific Alfa Aesar A17495.14

Request Bulk or Custom Format

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

3-Phenoxyaniline was used in the preparation of 4-phenoxyaniline-3?-bromopropionamide. It is also used in the design, synthesis and evaluation of potential inhibitors of HIV gp120-CD4 interactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenoxyaniline was used in the preparation of 4-phenoxyaniline-3′-bromopropionamide. It is also used in the design, synthesis and evaluation of potential inhibitors of HIV gp120-CD4 interactions.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3586-12-7
MDL Number MFCD00041891

Specifications

Melting Point 35°C to 37°C
Density 1.1583
Boiling Point 329°C to 330°C
Flash Point >110°C (230°F)
Molecular Formula C12H11NO
Refractive Index 1.1583
Quantity 25 g
Synonym benzenamine, 3-phenoxy, 3-aminodiphenyl ether, aniline, m-phenoxy, m-phenoxyaniline, m-aminophenyl phenyl ether, 3-phenoxyphenylamine, unii-k14xxp3gja, k14xxp3gja, 3-aminophenyl phenyl ether, 3-phenoxy aniline
Solubility Information Slightly soluble in water.
InChI Key UCSYVYFGMFODMY-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)N
IUPAC Name 3-phenoxyaniline
Molecular Weight (g/mol) 185.23
PubChem CID 77135
Formula Weight 185.23
Percent Purity 98%
Chemical Name or Material 3-Phenoxyaniline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249068 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog NumberA17495.14,A17495.06,A17495.22
Lot Number10238807 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberA17495.14,A17495.06,A17495.22
Lot Number10100150 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17495.14,A17495.06,A17495.22
Lot NumberJ5873A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17495.14,A17495.06,A17495.22
Lot Number10183246 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17495.14,A17495.06,A17495.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Phenoxyaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.