missing translation for 'onlineSavingsMsg'
Learn More
3'-Nitroacetanilide, 98+%, Thermo Scientific Chemicals Product Code.: 11419567

3'-Nitroacetanilide, 98+%, Thermo Scientific Chemicals

Product Code. 11419567
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11419567

Brand: Thermo Scientific Alfa Aesar B22062.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-28-1
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00017015
InChI Key KFTYNYHJHKCRKU-UHFFFAOYSA-N
Synonym n-3-nitrophenyl acetamide, 3'-nitroacetanilide, m-nitroacetanilide, 3-nitroacetanilide, acetamide, n-3-nitrophenyl, n-acetyl-m-nitroaniline, 3-nitro-n-acetylaniline, acetanilide, 3'-nitro, unii-qgh8s22nbp, qgh8s22nbpShow MoreShow Less
PubChem CID 31206
IUPAC Name N-(3-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 151°C to 153°C
Quantity 100 g
Beilstein 2211961
Merck Index 14,6581
Formula Weight 180.16
Percent Purity ≥98%
Chemical Name or Material 3'-Nitroacetanilide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10167320 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22062.06,B22062.14,B22062.22
Lot NumberL15N02 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22062.06,B22062.14,B22062.22
Lot Number10148417 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22062.06,B22062.14,B22062.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3'-Nitroacetanilide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.