missing translation for 'onlineSavingsMsg'
Learn More
3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 10551991

3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10551991
250 g, Plastic bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10551991

Brand: Thermo Scientific Acros 162012500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6276-54-6
Molecular Formula C3H8ClN·HCl
Molecular Weight (g/mol) 130.02
MDL Number MFCD00012913
InChI Key IHPRVZKJZGXTBQ-UHFFFAOYSA-N
Synonym 3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b
PubChem CID 11469095
IUPAC Name 3-chloropropan-1-amine;hydrochloride
SMILES C(CN)CCl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 148°C to 150°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Ionised Chloride) (Argentometry)
Packaging Plastic bottle
Linear Formula Cl(CH2)3NH2·HCl
Quantity 250 g
Beilstein 04,148
Solubility Information Solubility in water: freely soluble
Formula Weight 130.02
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Chloropropylamine hydrochloride, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460337 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number162010500,162012500,162010010,16201-25KG
Lot NumberA0432045 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number162010500,162012500,16201-100G,162010010,16201-25KG,16201-1KG
Lot NumberA0453082 Certificate TypeCertificate of Analysis Date16/05/2023 Catalog Number162010500,162012500,162010010
Lot NumberA0450774 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number162010500,162012500,162010010
Lot NumberA0400959 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number162010500,162012500,16201-100G,162010010,16201-25KG,16201-1KG
compliance-icons
Product Identifier
  • 3-Chloropropylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.