missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-5-fluorobenzoic acid, 97%, Thermo Scientific™ Product Code.: 15435447

3-Bromo-5-fluorobenzoic acid, 97%, Thermo Scientific™

Product Code. 15435447
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15435447

Brand: Thermo Scientific Alfa Aesar H32692.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Bromo-5-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176548-70-2
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD01569540
InChI Key KLSLJMGWUPAQGZ-UHFFFAOYSA-M
Synonym 5-bromo-3-fluorobenzoic acid, 3-fluoro-5-bromobenzoic acid, 3-bromo-5-fluorobenzoicacid, 5-fluoro-3-bromobenzoic acid, 3-bromo-5-fluoro-benzoic acid, 3-bromo-5-fluoro benzoic acid, buttpark 24\01-97, benzoic acid, 3-bromo-5-fluoro, pubchem2154, acmc-1c8tm
PubChem CID 2773339
IUPAC Name 3-bromo-5-fluorobenzoic acid
SMILES [O-]C(=O)C1=CC(F)=CC(Br)=C1

Specifications

Melting Point 141°C to 145°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 219.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-fluorobenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10162807 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10160983 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10178861 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10196256 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-5-fluorobenzoic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.