missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-4-nitropyridine N-oxide, 98+%, Thermo Scientific Chemicals Product Code.: 11318896

3-Bromo-4-nitropyridine N-oxide, 98+%, Thermo Scientific Chemicals

Product Code. 11318896
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11318896

Brand: Thermo Scientific Alfa Aesar L19524.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

3-Bromo-4-nitropyridine N-oxide is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-nitropyridine N-oxide is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Sparingly Soluble in water (0.92 g/L at 25°C).

Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1678-49-5
Molecular Formula C5H3BrN2O3
Molecular Weight (g/mol) 218.994
MDL Number MFCD00085925
InChI Key YZQMKADIENBVLH-UHFFFAOYSA-N
Synonym 3-bromo-4-nitropyridine n-oxide, 3-bromo-4-nitropyridine 1-oxide, 3-bromo-4-nitropyridin-1-ium-1-olate, pyridine, 3-bromo-4-nitro-, 1-oxide, 3-bromo-4-nitropyridine-n-oxide, 3-bromo-4-nitro-1-oxido-pyridin-1-ium, 3-bromo-4-nitropyridin-1-ol, acmc-1bxbw, 3-bromo-4-nitropyridine oxide, 3-bromo-4-nitropyridin-n-oxideShow MoreShow Less
PubChem CID 548521
IUPAC Name 3-bromo-4-nitro-1-oxidopyridin-1-ium
SMILES C1=C[N+](=CC(=C1[N+](=O)[O-])Br)[O-]

Specifications

Melting Point 152°C to 154°C
Quantity 1 g
Beilstein 150822
Solubility Information Sparingly Soluble in water (0.92g/L at 25°C).
Formula Weight 219
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-4-nitropyridine N-oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242557 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog NumberL19524.06,L19524.03,L19524.14
Lot Number10155554 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL19524.06,L19524.03,L19524.14
Lot Number10236210 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL19524.06,L19524.03,L19524.14
Lot Number10176981 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL19524.06,L19524.03,L19524.14
Lot Number10163679 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL19524.06,L19524.03,L19524.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-4-nitropyridine N-oxide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.