missing translation for 'onlineSavingsMsg'
Learn More
3,5-Dihydroxybenzyl alcohol, 98%, Thermo Scientific™ Product Code.: 11400788

3,5-Dihydroxybenzyl alcohol, 98%, Thermo Scientific™

Product Code. 11400788
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11400788

Brand: Thermo Scientific Alfa Aesar B25248.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dihydroxybenzyl alcohol may be used as monomer for the synthesis of a series of monodisperse dendritic polyether macromolecules. It may be employed in the synthesis of dimethylsilyl linked dihydroxybenzyl alcohol based dendrimers.

Solubility
Soluble in DMSO, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29654-55-5
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00016867
InChI Key NGYYFWGABVVEPL-UHFFFAOYSA-N
Synonym 3,5-dihydroxybenzyl alcohol, 5-hydroxymethyl-1,3-benzenediol, 5-hydroxymethyl benzene-1,3-diol, 5-hydroxymethyl resorcinol, benzyl alcohol, 3,5-dihydroxy, 1,3-benzenediol, 5-hydroxymethyl, 3,5-dihydroxybenzylalcohol, 3,5-dihydroxy benzyl alcohol, pubchem7418, acmc-1cjogShow MoreShow Less
PubChem CID 34661
IUPAC Name 5-(hydroxymethyl)benzene-1,3-diol
SMILES C1=C(C=C(C=C1O)O)CO

Specifications

Melting Point 182°C to 186°C (decomposition)
Quantity 1 g
Solubility Information Soluble in DMSO,methanol.
Formula Weight 140.14
Percent Purity 98%
Chemical Name or Material 3,5-Dihydroxybenzyl alcohol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH09Z014 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10183713 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10194714 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10159554 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10179741 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Dihydroxybenzyl alcohol, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.