Learn More
3,4-Dimethoxyphenol, 98%, Thermo Scientific Chemicals
Description
3,4-Dimethoxyphenol is used in the preparation of 5,6-dimethoxy benzofuranone derivatives and multi-target anti Alzheimer compounds. It is also used in the preparation of 3,4-dimethoxyphenyl-beta-D-glucopyranoside and 4-(but-2-enyloxy)-1,2-dimethoxybenzene. Further, it serves as a precursor for 4H-chromenes synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3,4-Dimethoxyphenol is used in the preparation of 5,6-dimethoxy benzofuranone derivatives and multi-target anti Alzheimer compounds. It is also used in the preparation of 3,4-dimethoxyphenyl-beta-D-glucopyranoside and 4-(but-2-enyloxy)-1,2-dimethoxybenzene. Further, it serves as a precursor for 4H-chromenes synthesis.
Solubility
Soluble in water.
Notes
Light sensitive. Store in a cool place. Incompatible with bases, acid chlorides, acid anhydrides and oxidizing agents.

Specifications
Specifications
Melting Point | 79°C to 82°C |
Boiling Point | 167°C (14 mmHg) |
Quantity | 25 g |
Beilstein | 1366650 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 154.16 |
Percent Purity | 98% |
Chemical Name or Material | 3,4-Dimethoxyphenol |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10222668 | Certificate of Analysis | 30/01/2025 | A11344.06,A11344.14,A11344.22 |
10217324 | Certificate of Analysis | 22/11/2021 | A11344.06,A11344.14,A11344.22 |
10107568 | Certificate of Analysis | 22/11/2021 | A11344.06,A11344.14,A11344.22 |
10136155 | Certificate of Analysis | 22/11/2021 | A11344.06,A11344.14,A11344.22 |
10107351 | Certificate of Analysis | 22/11/2021 | A11344.06,A11344.14,A11344.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.