Learn More
Fisher Chemical 25KG SULFAMIC ACID - SPECIFIED REAGENT
Brand: Fisher Chemical S/8640/70
391.76 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
25KG SULFAMIC ACID - SPECIFIED REAGENT
Description
Safety and Handling
- Sulfamic acid
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2534783 | Certificate of Analysis | 18/04/2025 | S/8640/70 |