missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 250 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
BIOPHARMAGREENER_CHOICE

For use with Agilent/Velocity11 automation systems

62.84€ - 474.70€

Specifications

Volume (Metric) 250 μL
Filtered Non-filtered
Color Clear
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
View More Specs

Products 9
Product Code Brand Sterility Tip Style Features Price Quantity & Availability  
Product Code Brand Sterility Tip Style Features Price Quantity & Availability  
12555567
GREENER_CHOICE
View Documents
Axygen™
VT-250-R
Non-sterile Standard Non-pyrogenic This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
10313370
BIOPHARMAGREENER_CHOICE
View Documents Promotion Details
Axygen™
VT-250-R
Non-sterile Standard Non-pyrogenic
385.40€
Pack of 4800
Estimated Shipment: 26-08-2025
to see stock.
Add to basket
 
12786649
GREENER_CHOICE
View Documents
Axygen™
VT-250-L-R
Non-sterile Low Retention Non-pyrogenic This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
12535567
GREENER_CHOICE
View Documents
Axygen™
VT-250-L-R
Non-sterile Low Retention Non-pyrogenic This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
12585567
GREENER_CHOICE
View Documents
Axygen™
VT-250-R-S
Sterile Standard Non-pyrogenic This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
10221480
BIOPHARMAGREENER_CHOICE
View Documents
Axygen™
VT-250-R-S
Sterile Standard Non-pyrogenic
474.70€
Pack of 4800
Estimated Shipment: 26-08-2025
to see stock.
Add to basket
 
12716659
BIOPHARMAGREENER_CHOICE
View Documents
Axygen™
VT-250-R-LAB
Non-sterile Standard Non-pyrogenic, Bottom Label
423.64€
Pack of 4800
In Stock
to see stock.
Add to basket
 
12565567
GREENER_CHOICE
View Documents
Axygen™
VT-250-R-LAB
Non-sterile Standard Non-pyrogenic, Bottom Label This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
12575567
View Documents
Axygen™
VT-250-R-LAB-S
Sterile Standard Non-pyrogenic, Bottom Label This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description

Description

Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

  • Clear polypropylene
  • Maxymum Recovery™ improves accuracy and precision while reducing sample loss
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Available in nonsterile or sterile

Specifications

Specifications

250 μL
Clear
DNase, Rnase Free
Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT
Non-filtered
ANSI
Rack
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ 250 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.