missing translation for 'onlineSavingsMsg'
Learn More
2-Fluoro-5-methylaniline, 98%, Thermo Scientific Chemicals Product Code.: 11303047

2-Fluoro-5-methylaniline, 98%, Thermo Scientific Chemicals

Product Code. 11303047
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11303047

Brand: Thermo Scientific Alfa Aesar L13411.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-84-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007654
InChI Key QZUXMXZNVAJNSE-UHFFFAOYSA-N
Synonym 3-amino-4-fluorotoluene, 6-fluoro-m-toluidine, 2-fluoro-5-methylphenylamine, 2-fluoro-5-methylbenzenamine, benzenamine, 2-fluoro-5-methyl, 2-fluoro-5-methylbenzeneamine, 2-fluoro-5-methyl-aniline, pubchem1507, 2-fluoro-5-metylaniline, acmc-209k1v
PubChem CID 262970
IUPAC Name 2-fluoro-5-methylaniline
SMILES CC1=CC=C(F)C(N)=C1

Specifications

Density 1.109
Boiling Point 80°C to 86°C (11 mmHg)
Flash Point 81°C (177°F)
Odor Characteristic
Refractive Index 1.533
Quantity 25 g
UN Number UN2810
Beilstein 2715995
Formula Weight 125.15
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-methylaniline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10167863 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13411.06,L13411.14
Lot NumberK19W057 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13411.06,L13411.14
Lot Number10160827 Certificate TypeCertificate of Analysis Date02/12/2021 Catalog NumberL13411.06,L13411.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluoro-5-methylaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.