Learn More
2-Amino-2',5-dichlorobenzophenone, 98%, Thermo Scientific Chemicals
Description
2-Amino-2',5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Amino-2′,5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Specifications
Specifications
Melting Point | 87°C to 89°C |
Odor | Odorless |
Quantity | 10 g |
Beilstein | 652774 |
Solubility Information | Insoluble in water. |
Formula Weight | 266.13 |
Percent Purity | 99% |
Chemical Name or Material | 2-Amino-2',5-dichlorobenzophenone |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.