missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylnaphthalene, 99%, Thermo Scientific Chemicals

Product Code. 11422527
1000 g
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.

Solubility
Soluble in ethanol, acetone and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-08-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00004108
InChI Key XSAYZAUNJMRRIR-UHFFFAOYSA-N
Synonym 2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
PubChem CID 7122
ChEBI CHEBI:52364
IUPAC Name 1-naphthalen-2-ylethanone
SMILES CC(=O)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 52°C to 56°C
Boiling Point 300°C to 301°C
Flash Point 168°C (334°F)
Quantity 1000 g
UN Number UN3077
Beilstein 774965
Solubility Information Soluble in ethanol,acetone and ether. Insoluble in water.
Formula Weight 170.21
Percent Purity 99%
Chemical Name or Material 2-Acetylnaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Acetylnaphthalene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.