missing translation for 'onlineSavingsMsg'
Learn More
2'-Acetonaphthone, 99%, Thermo Scientific Chemicals Product Code.: 10397520

2'-Acetonaphthone, 99%, Thermo Scientific Chemicals

Product Code. 10397520
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 10397520

Brand: Thermo Scientific Acros 158885000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-08-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00004108
InChI Key XSAYZAUNJMRRIR-UHFFFAOYSA-N
Synonym 2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketoneShow MoreShow Less
PubChem CID 7122
ChEBI CHEBI:52364
IUPAC Name 1-naphthalen-2-ylethanone
SMILES CC(=O)C1=CC2=CC=CC=C2C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53.0°C to 56.0°C
Color White
Boiling Point 300.0°C to 301.0°C
Flash Point 168°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C10H7COCH3
Quantity 500 g
Beilstein 07, 402
Solubility Information Solubility in water: insoluble
Formula Weight 170.21
Percent Purity 99%
Physical Form Fine Crystalline Powder and Chunks
Chemical Name or Material 2'-Acetonaphthone, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463899 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number158881000,158885000
Lot NumberA0463466 Certificate TypeCertificate of Analysis Date10/06/2024 Catalog Number158881000,158885000
Lot NumberA0448586 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number158881000,158885000
Lot NumberA0441396 Certificate TypeCertificate of Analysis Date21/04/2022 Catalog Number158881000,158885000
Lot NumberA0382451 Certificate TypeCertificate of Analysis Date24/03/2022 Catalog Number158881000,158885000
compliance-icons compliance-icons
Product Identifier
  • 2'-Acetonaphthone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2'-Acetonaphthone, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.