Learn More
2-(4-Ethoxyphenyl)ethanol, 98%, Thermo Scientific Chemicals Product Code.: 11307505
2-(4-Ethoxyphenyl)ethanol, 98%, Thermo Scientific Chemicals
Product Code. 11307505
10 g
Click to view available options
Quantity:
2 g
10 g
Product Code. 11307505
Brand: Thermo Scientific Alfa Aesar L05597.09
Request Bulk or Custom Format
2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22545-15-9
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00016570
InChI Key CNMVSNTVPZWQMI-UHFFFAOYSA-N
Synonym 2-4-ethoxyphenyl ethanol, p-ethoxyphenethyl alcohol, 4-2-hydroxyethyl phenetol, 4-ethoxybenzeneethanol, 4-ethoxy-benzeneethanol, 4-ethoxyphenethyl alcohol, benzeneethanol,4-ethoxy, acmc-1cit4, benzeneethanol, 4-ethoxy, 2-4-ethoxyphenyl ethan-1-olShow MoreShow Less
PubChem CID 89752
IUPAC Name 2-(4-ethoxyphenyl)ethanol
SMILES CCOC1=CC=C(CCO)C=C1
Specifications
Melting Point 40°C to 44°C
Boiling Point 138°C to 141°C (8 mmHg)
Quantity 10 g
Beilstein 3239377
Solubility Information Soluble in water.
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 2-(4-Ethoxyphenyl)ethanol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10115963 Certificate TypeCertificate of Analysis Date02/11/2023 Catalog NumberL05597.04,L05597.09
Lot Number10115936 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL05597.04,L05597.09
Lot Number10115963 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL05597.04,L05597.09
Lot NumberGBFA022762 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL05597.04,L05597.09
RUO – Research Use Only
Product Content Correction
Product Title
2-(4-Ethoxyphenyl)ethanol, 98%, Thermo Scientific Chemicals >