missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dimethylthiophenol, 95%, Thermo Scientific Chemicals Product Code.: 11450747

2,4-Dimethylthiophenol, 95%, Thermo Scientific Chemicals

Product Code. 11450747
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11450747

Brand: Thermo Scientific Alfa Aesar B23161.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13616-82-5
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010019
InChI Key AMNLXDDJGGTIPL-UHFFFAOYSA-N
Synonym 2,4-dimethylthiophenol, 2,4-dimethyl thiophenol, benzenethiol, 2,4-dimethyl, 2,4-thioxylenol, 2,4-xylenethiol, 4-mercapto-m-xylene, 2,4-dimethylbenzene-1-thiol, 4-methylthiocresol, 4-thio-m-xylene, m-xylene-4-thiolShow MoreShow Less
PubChem CID 83617
IUPAC Name 2,4-dimethylbenzenethiol
SMILES CC1=CC(=C(C=C1)S)C

Specifications

Density 1.02
Boiling Point 207°C to 208°C
Flash Point 87°C (189°F)
Odor Unpleasant
Refractive Index 1.569
Quantity 25 g
UN Number UN2810
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 95%
Chemical Name or Material 2,4-Dimethylthiophenol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251706 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB23161.06,B23161.14,B23161.22
Lot Number10200028 Certificate TypeCertificate of Analysis Date11/09/2023 Catalog NumberB23161.06,B23161.14,B23161.22
Lot Number10244655 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog NumberB23161.06,B23161.14,B23161.22
Lot Number10200028 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23161.06,B23161.14,B23161.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dimethylthiophenol, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.