Learn More
2,4-dichlorotoluene, Mettler Toledo™
Shop All Mettler Toledo Products
Click to view available options
Quantity:
9 mL
Specifications
Specifications
Description | 2,4-dichlorotoluene |
Quantity | 9 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Promotions
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Product Title
2,4-dichlorotoluene, Mettler Toledo™ >