missing translation for 'onlineSavingsMsg'
Learn More
(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 15490116

(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15490116
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15490116

Brand: Thermo Scientific Alfa Aesar H52779.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24716-93-6
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD01318270
InChI Key USQHEVWOPJDAAX-WDSKDSINSA-N
PubChem CID 7128323
IUPAC Name (1S,2S)-2-aminocyclohexane-1-carboxylic acid
SMILES C1CCC(C(C1)C(=O)O)N

Specifications

Synonym 1s,2s-2-aminocyclohexanecarboxylic acid, 1s,2s-2-aminocyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 2-amino-, 1s,2s, 1s,2s-2-aminocyclohexanecarboxylicacid, 1s,2s-2-amino-cyclohexanecarboxylic acid, 2beta-amino-1alpha-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid,2-amino-, 1s,2s, 24716-92-5 1s,2s-2-aminocyclohexane-1-carboxylic acid
Solubility Information Soluble in water.
Quantity 250 mg
Formula Weight 143.19
Optical Rotation +77° (c=1 in Water)
Percent Purity 97%
Chemical Name or Material (1S,2S)-2-Aminocyclohexanecarboxylic acid
compliance-icons
Product Identifier
  • (1S,2S)-2-Aminocyclohexanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : (1S,2S)-2-Aminocyclohexanecarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.