Learn More
Thermo Scientific Alfa Aesar 100ML THF, anh., 99.8+%, BHT-free, over mol. sieves,
Brand: Thermo Scientific Alfa Aesar 047327.AE
49.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
100ML THF, anh., 99.8+%, BHT-free, over mol. sieves,
Description
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2533812 | Certificate of Analysis | 10/04/2025 | 047327.AE,047327.K2 |
J30Q733 | Certificate of Analysis | 02/04/2025 | 047327.AE,047327.K2 |
2528720 | Certificate of Analysis | 27/02/2025 | 047327.AE,047327.K2 |
2528719 | Certificate of Analysis | 27/02/2025 | 047327.AE,047327.K2 |
2528718 | Certificate of Analysis | 27/02/2025 | 047327.AE,047327.K2 |