Learn More
Keystone Cleanroom Products 1421B Tyvek™ Autoclave Bag
1421B Tyvek™ Autoclave Bag designed for the steam sterilization of large, or odd shaped pharmaceutical equipment change parts.
1861.60€ - 5350.40€
Specifications
Product Type | Autoclave Bag |
---|---|
Autoclavable | Autoclavable |
Certifications/Compliance | Certificate of Conformance (COC) |
Color | White |
Disposable | Single-use |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
30020294
|
Keystone Cleanroom Products
6STK09-512 |
9.5 x 12 in. |
1861.60€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020126
|
Keystone Cleanroom Products
6STK0212 |
12 x 2 in. |
2677.44€
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020072
|
Keystone Cleanroom Products
6STK1212 |
12 x 12 in. |
2321.37€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020064
|
Keystone Cleanroom Products
6STK1015 |
15 x 10 in. |
2082.46€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020078
|
Keystone Cleanroom Products
6STK1216 |
16 x 12 in. |
2379.96€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020106
|
Keystone Cleanroom Products
6STK1218 |
18 x 12 in. |
2510.67€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020077
|
Keystone Cleanroom Products
6STK1418 |
18 x 14 in. |
2605.33€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020090
|
Keystone Cleanroom Products
6STK2020 |
20 x 20 in. |
2866.77€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020336
|
Keystone Cleanroom Products
6STK1624 |
24 x 16 in. |
2812.67€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020250
|
Keystone Cleanroom Products
6STK2024 |
24 x 20 in. |
2902.83€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020168
|
Keystone Cleanroom Products
6STK2424 |
24 x 24 in. |
2880.29€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020130
|
Keystone Cleanroom Products
6STK2426 |
26 x 24 in. |
3177.79€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020373
|
Keystone Cleanroom Products
6STK1227 |
27 x 12 in. |
2947.90€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020345
|
Keystone Cleanroom Products
6STK2844 |
28 x 44 in. |
4539.04€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020346
|
Keystone Cleanroom Products
6STK1030 |
30 x 10 in. |
2799.15€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020110
|
Keystone Cleanroom Products
6STK1830 |
30 x 18 in. |
2902.83€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020351
|
Keystone Cleanroom Products
6STK2030 |
30 x 20 in. |
2902.83€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020344
|
Keystone Cleanroom Products
6STK1232 |
32 x 12 in. |
2222.19€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020053
|
Keystone Cleanroom Products
6STK2436 |
36 x 24 in. |
3394.13€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30019999
|
Keystone Cleanroom Products
6STK3036 |
36 x 30 in. |
3831.37€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020260
|
Keystone Cleanroom Products
6STK3636 |
36 x 36 in. |
4408.33€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020023
|
Keystone Cleanroom Products
6STK1240 |
40 x 12 in. |
3416.67€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020049
|
Keystone Cleanroom Products
6STK2040 |
40 x 20 in. |
2902.83€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020316
|
Keystone Cleanroom Products
6STK2442 |
42 x 24 in. |
3817.85€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020267
|
Keystone Cleanroom Products
6STK4242 |
42 x 42 in. |
2418.27€
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020359
|
Keystone Cleanroom Products
6STK1246 |
46 x 12 in. |
3105.65€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020032
|
Keystone Cleanroom Products
6STK1248 |
48 x 12 in. |
3191.31€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020322
|
Keystone Cleanroom Products
6STK2448 |
48 x 24 in. |
4237.04€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020282
|
Keystone Cleanroom Products
6STK3648 |
48 x 36 in. |
2483.63€
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020206
|
Keystone Cleanroom Products
6STK4848 |
48 x 48 in. |
2587.31€
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020246
|
Keystone Cleanroom Products
6STK3250 |
50 x 32 in. |
5224.19€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020269
|
Keystone Cleanroom Products
6STK5054 |
54 x 50 in. |
2457.48€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020157
|
Keystone Cleanroom Products
6STK3660 |
60 x 36 in. |
2659.42€
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020043
|
Keystone Cleanroom Products
6STK1062 |
62 x 10 in. |
3349.08€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30020079
|
Keystone Cleanroom Products
6STK2462 |
62 x 24 in. |
5350.40€
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Tyvek™ on both sides for maximum breathability during sterilization
- Eliminates need to cut material and the use of autoclave tape
- Fast and consistent for equipment preparation area
- Creates secondary level of protection during aseptic presentation of equipment
- Sealed seams eliminate sewing threads and greatly reduce particulates
- Resists tearing and punctures
- Superior microbial barrier
- Lint-free Category 1 (Helmke Drum)
- Manufactured and packaged in a ISO 6 cleanroom environment
- Certificates of Conformance sent with shipment
Specifications
Autoclave Bag | |
Certificate of Conformance (COC) | |
Single-use | |
Pharmaceutical Equipment and Components | |
Non-sterile | |
Up to 250°F | |
5.3 mil |
Autoclavable | |
White | |
Steam Sterilization | |
Tyvek | |
0.13 mm | |
Up to 125°C |