missing translation for 'onlineSavingsMsg'
Learn More
1-Phenoxy-2-propanol, tech. 85%, Thermo Scientific Chemicals Product Code.: 11341637

1-Phenoxy-2-propanol, tech. 85%, Thermo Scientific Chemicals

Product Code. 11341637
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11341637

Brand: Thermo Scientific Alfa Aesar L04596.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 770-35-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00016861
InChI Key IBLKWZIFZMJLFL-UHFFFAOYNA-N
Synonym 1-phenoxy-2-propanol, 2-propanol, 1-phenoxy, phenoxyisopropanol, propylene phenoxetol, 2-phenoxy-1-methylethanol, 1-phenoxy-propan-2-ol, phenyl-.beta.-hydroxypropyl ether, dsstox_cid_7312, dsstox_rid_78402, dsstox_gsid_27312Show MoreShow Less
PubChem CID 92839
IUPAC Name 1-phenoxypropan-2-ol
SMILES CC(O)COC1=CC=CC=C1

Specifications

Density 1.06
Boiling Point 242°C to 244°C
Flash Point 120°C (248°F)
Refractive Index 1.523
Quantity 250 g
Beilstein 1941356
Formula Weight 152.19
Percent Purity 85%
Grade Technical
Chemical Name or Material 1-Phenoxy-2-propanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251174 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL04596.30,L04596.18
Lot Number10237396 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL04596.30,L04596.18
Lot NumberK22W039 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL04596.30,L04596.18
Lot Number10229624 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL04596.30,L04596.18
Lot Number10143469 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL04596.30,L04596.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Phenoxy-2-propanol, tech. 85%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.