missing translation for 'onlineSavingsMsg'
Learn More
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis, Thermo Scientific Chemicals Product Code.: 10113781

1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis, Thermo Scientific Chemicals

Product Code. 10113781
2.5 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10113781

Brand: Thermo Scientific Acros 354900025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point -24.0°C
Density 1.0300g/mL
Boiling Point 202.0°C
Quantity 2.5 L
Flash Point 91°C
Packaging Glass Bottle
Refractive Index 1.4690 to 1.4710
Assay Percent Range 99.4% min. (GC)
Specific Gravity 1.03
Viscosity 1.67 mPa.s (20°C)
Beilstein 21,237
Water 0.1% max. (K.F.)
Fieser 01,696; 02,281; 09,316; 11,346
Merck Index 15,619
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 99.13
Percent Purity 99.50%
Grade Peptide Synthesis
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467044 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog Number354900100,354900010,354900025
Lot NumberA0460100 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number354900100,354900010,354900025
Lot NumberA0399416 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number354900100,354900010,354900025
Lot NumberA0407716 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number354900100,354900010,354900025
Lot NumberA0445034 Certificate TypeCertificate of Analysis Date12/08/2022 Catalog Number354900100,354900010,354900025
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.