missing translation for 'onlineSavingsMsg'
Learn More
1-Fluoro-4-iodobenzene, 99%, Thermo Scientific Chemicals Product Code.: 11434064

1-Fluoro-4-iodobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11434064
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11434064

Brand: Thermo Scientific Alfa Aesar A12008.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Fluoroiodobenzene was used in the preparation of fluorotriphenylene derivatives. 4-[F] Fluoroiodobenzene is a versatile building block in 18F radiochemistry and is used in various transition metal-mediated C-C and C-N cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoroiodobenzene was used in the preparation of fluorotriphenylene derivatives. 4-[F] Fluoroiodobenzene is a versatile building block in 18F radiochemistry and is used in various transition metal-mediated C-C and C-N cross-coupling reactions.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool dry place. Keep container tightly sealed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-34-1
Molecular Formula C6H4FI
Molecular Weight (g/mol) 222.00
MDL Number MFCD00001052
InChI Key KGNQDBQYEBMPFZ-UHFFFAOYSA-N
Synonym 4-fluoroiodobenzene, p-fluoroiodobenzene, benzene, 1-fluoro-4-iodo, 4-iodofluorobenzene, p-iodofluorobenzene, 4-fluoro-1-iodobenzene, 4-fluoro-iodobenzene, 1-fluoro-4-iodo-benzene, 1-iodo-4-fluorobenzene, 4-fluroiodobenzene
PubChem CID 9605
IUPAC Name 1-fluoro-4-iodobenzene
SMILES FC1=CC=C(I)C=C1

Specifications

Melting Point -20°C
Density 1.925
Boiling Point 182°C to 184°C
Flash Point 68°C (154°F)
Refractive Index 1.583
Quantity 50 g
Beilstein 1853970
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 222
Percent Purity 99%
Chemical Name or Material 1-Fluoro-4-iodobenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Fluoro-4-iodobenzene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.