missing translation for 'onlineSavingsMsg'
Learn More
1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals Product Code.: 10727202

1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals

Product Code. 10727202
50 g, Glass bottle
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10727202

Brand: Thermo Scientific Acros 249460500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-41-8
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001033
InChI Key MPEOPBCQHNWNFB-UHFFFAOYSA-N
Synonym 2-chloroiodobenzene, o-chloroiodobenzene, o-iodochlorobenzene, 2-iodochlorobenzene, benzene, 1-chloro-2-iodo, 2-chlorophenyl iodide, 2-chloro-1-iodobenzene, 1-chloro-2-iodo-benzene, chlorjodbenzol, iodochlorobenzeneShow MoreShow Less
PubChem CID 11993
IUPAC Name 1-chloro-2-iodobenzene
SMILES ClC1=CC=CC=C1I

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 1.0°C
Color Yellow
Density 1.9500g/mL
Boiling Point 234.0°C to 235.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClC6H4I
Refractive Index 1.6330 to 1.6350
Quantity 50 g
Beilstein 05, 220
Specific Gravity 1.95
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 238.46
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-iodobenzene, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0433285 Certificate TypeCertificate of Analysis Date02/09/2024 Catalog Number249460500,249460050
Lot NumberA0433285 Certificate TypeCertificate of Analysis Date03/09/2021 Catalog Number
Lot NumberA0372748 Certificate TypeCertificate of Analysis Date16/11/2020 Catalog Number249460500,249460050
Lot NumberA0331546 Certificate TypeCertificate of Analysis Date26/08/2020 Catalog Number249460500,249460050
compliance-icons
Product Identifier
  • 1-Chloro-2-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.